Intravenøst kateter, Surflash™
Intravenøst kateter, G 18, grøn, PU, 32 mm
Katalog # TERUSR-FF1832
Leverandør: TERUMO
Gå til:
Product Details & Documents
Surflash is made of PU, which is firm at room temperature but soft at body temperature, making the catheter more comfortable.
- Only exists in the straight version
- Enables quick and easy confirmation of the placement in the vein by an additional flashback
- Higher certainty of proper placement
Helpful with patients whose veins are hard to detect and in which venipuncture is more difficult, such as small children, people undergoing chemotherapy.
Documents
Specifikationer
- Måler:G 18
- Farve:grøn
- Beskrivelse:Intravenøst kateter
- Længde:32 mm
- Materiale:PU
- Seq:003