Glove liners, BCR®
Full-finger cleanroom liners, green cuff, Full-finger cleanroom liners, green cuff, S, hvid
Katalog # BERKBGL7.200.SB
Leverandør: BERKSHIRE
Gå til:
Product Details & Documents
These single knit glove liners are made out of high quality 100% continuous filament pre-shrunk nylon. They are lightweight, soft, and comfortable to wear. Available as half-finger and full-finger versions.
- Protect against skin irritation caused by latex or PVC gloves
- Constructed using special seamless techniques
- Ambidextrous for easy stock control
- Launderable for multiple reuse
- Colour-coded cuffs to identify sizes
Suitable for use in cleanrooms.
Specifikationer
- Handske størrelse:S
- Farve:hvid
- Beskrivelse:Full-finger cleanroom liners, green cuff
- Seq:03
- Model:Full-finger cleanroom liners, green cuff
- Cat. No.:BERKBGL7.200.SB
- Ambidextrous/Hånd-specifik:Ambidextrous
- Engangs/Genbrugelig:Genanvendelig