Orders LinkContactUsLinkComponent
Glove liners, BCR®
Glove liners, BCR®
Full-finger cleanroom liners, green cuff, Full-finger cleanroom liners, green cuff, S, hvid
Katalog # BERKBGL7.200.SB
Leverandør:   BERKSHIRE
Glove liners, BCR®
Katalog # BERKBGL7.200.SB
Leverandør:  BERKSHIRE
Glove liners, BCR®
Glove liners, BCR®
Full-finger cleanroom liners, green cuff, Full-finger cleanroom liners, green cuff, S, hvid
Price
4.590,00 kr
Log ind for tilgængelighed
1 * 200 Par
Product Details & Documents
These single knit glove liners are made out of high quality 100% continuous filament pre-shrunk nylon. They are lightweight, soft, and comfortable to wear. Available as half-finger and full-finger versions.
  • Protect against skin irritation caused by latex or PVC gloves
  • Constructed using special seamless techniques
  • Ambidextrous for easy stock control
  • Launderable for multiple reuse
  • Colour-coded cuffs to identify sizes
Suitable for use in cleanrooms.
Specifikationer
  • Handske størrelse:
    S
  • Farve:
    hvid
  • Beskrivelse:
    Full-finger cleanroom liners, green cuff
  • Seq:
    03
  • Model:
    Full-finger cleanroom liners, green cuff
  • Cat. No.:
    BERKBGL7.200.SB
  • Ambidextrous/Hånd-specifik:
    Ambidextrous
  • Engangs/Genbrugelig:
    Genanvendelig