High visibility jacket, Strada 2 High-Visibility
orange/blå, L
Katalog # 113-0914
Leverandør: DELTA PLUS
Gå til:
Product Details & Documents
This high-visibility safety jacket (class 3) is made from high visibility, fluorescent polyester with internal polyurethane coating and polyester lining. The jacket includes a zip with press stud cover, two pockets with press stud pocket covers, two side pockets, an inside pocket, and waterproof seams. Suitable for ∼50 washes at 40 °C.
- Two reflective strips on each sleeve and strips on the body and shoulders
- Non detachable hood with velcro fastening, which can be folded in the collar
- Straight sleeve openings with wind-resistant, internal elastic cuffs
- Ventilation eyelets in the underarm area
- Can be tightened at the waist with elastic cord and toggle
Colour: Fluorescent yellow/marine blue or fluorescent orange/marine blue.
Certifications: EN 340; EN 342; EN 343; EN 471
Specifikationer
- Tøj størrelse:L
- Farve:orange/blå
- Seq:01
- Køn:Unisex
- Cat. No.:113-0914
- Sleeve or leg length:Long
- Risk area:HiVis
- Outfit applikationer:Industrial
- Lukkestørrelse:Med lynlås
- Stof:Polyester with internal polyurethane coating