Orders LinkContactUsLinkComponent
Accessories for SI Analytics titrators
Accessories for SI Analytics titrators
Micro-double platinum combination electrode for Karl-Fischer titrations, with fixed cable, double platinum pin and tapper NS 7,5 for coulometric titration vessels TZ 1751 + TZ 1754 (for use with TitroLine® KF / TitroLine® KF trace)
Katalog # SCOI285102060
Leverandør:   SI Analytics
Accessories for SI Analytics titrators
Katalog # SCOI285102060
Leverandør:  SI Analytics
Accessories for SI Analytics titrators
Micro-double platinum combination electrode for Karl-Fischer titrations, with fixed cable, double platinum pin and tapper NS 7,5 for coulometric titration vessels TZ 1751 + TZ 1754 (for use with TitroLine® KF / TitroLine® KF trace)
Price
3.210,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Micro-double platinum combination electrode for Karl-Fischer titrations, with fixed cable, double platinum pin and tapper NS 7,5 for coulometric titration vessels TZ 1751 + TZ 1754 (for use with TitroLine® KF / TitroLine® KF trace)
  • Seq:
    02