Accessories for SI Analytics titrators
Micro-double platinum combination electrode for Karl-Fischer titrations, with fixed cable, double platinum pin and tapper NS 7,5 for coulometric titration vessels TZ 1751 + TZ 1754 (for use with TitroLine® KF / TitroLine® KF trace)
Katalog # SCOI285102060
Leverandør: SI Analytics
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Micro-double platinum combination electrode for Karl-Fischer titrations, with fixed cable, double platinum pin and tapper NS 7,5 for coulometric titration vessels TZ 1751 + TZ 1754 (for use with TitroLine® KF / TitroLine® KF trace)
- Seq:02