Orders LinkContactUsLinkComponent
Ampicillin, natriumsalt
Glass bottle for solids, 250 g
  A6352.0250
Ampicillin, natriumsalt
  A6352.0250
 :  69-52-3
Ampicillin, natriumsalt
Glass bottle for solids, 250 g
 :  69-52-3
11.600,00 kr
1 * 250 g
 :  D(-)-α-Aminobenzylpenicillin Sodium Salt, Ampicillin Na, Ampicillin sodium salt, D-(−)-α-Aminobenzylpenicillin sodium salt Ampicillin sodium salt
  •  :69-52-3
  •  :C₁₆H₁₈N₃NaO₄S
  •  :MFCD00064313
  •  :371.39 g/mol
  •  :Køleskab
  •  :200-708-1
  • Pakning:
    Glass bottle for solids
  • Pakn:
    250 g