Cream butyrometer, without stopper
Butyrometer for cream, Roeder´s, 0 - 30 - 55 % : 0,5
Katalog # 614-0108
Leverandør: GERBER FUNKE
Gå til:
Product Details & Documents
According to Roeder´s weighing method.
Specifikationer
- Beskrivelse:Butyrometer for cream, Roeder´s, 0 - 30 - 55 % : 0,5
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Weighing boat for butter for butyrometers acc. to Roeder's methodWeighing boat for butter for butyrometers acc. to Roeder's method614-2034Log ind for tilgængelighed1 * 1 Items - 47,80 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Cream beaker, unperforated, for ice cream and condensed milk butyrometer and cream butyrometer acc. to Roeder´s methodCream beaker, unperforated, for ice cream and condensed milk butyrometer and cream butyrometer acc. to Roeder´s method614-2032Log ind for tilgængelighed1 * 1 Items - 18,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Stopper, rubber, with hole, for all weighing method butyrometersStopper, rubber, with hole, for all weighing method butyrometers614-2030Log ind for tilgængelighed1 * 100 Items - 540,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Stopper, rubber, for sealing the bulbs of all weighing method butyrometersStopper, rubber, for sealing the bulbs of all weighing method butyrometers614-2029Log ind for tilgængelighed1 * 100 Items - 180,00 kr