Orders LinkContactUsLinkComponent
Natriumdeoxycholat
Natriumdeoxycholat
Glass bottle for solids, 25 g
Katalog # PIER89905
Irriterende
Leverandør:   Thermo Fisher Scientific
Natriumdeoxycholat
Katalog # PIER89905
CAS nummer:  302-95-4
Natriumdeoxycholat
Natriumdeoxycholat
Glass bottle for solids, 25 g
CAS nummer:  302-95-4
Price
5.080,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
Synonymer:  Deoxycholic acid sodium salt, 3α,12α-Dihydroxy-5β-cholan-24-oic acid sodium salt, Sodium 3α,12α-dihydroxy-5β-cholanoate
  • CAS nummer:302-95-4
  • Formel:C₂₄H₃₉NaO₄
  • MDL nummer:MFCD00064139
  • Molecular Weight:414.56 g/mol
  • Smeltende Pt:357…365 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:206-132-7
Documents
Specifikationer
Specifikationer
  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g