Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
HyClone Cell Boost™ 2 Supplement

HyClone Cell Boost™ 2 Supplement

Cell Boost™ 2, Powder, 100 g, 100 g
Katalog # SH30596.01
HyClone Cell Boost™ 2 Supplement
Katalog # SH30596.01
CAS nummer:  
HyClone Cell Boost™ 2 Supplement
HyClone Cell Boost™ 2 Supplement

Cell Boost™ 2, Powder, 100 g, 100 g

CAS nummer:  
Price
1.840,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Supplements cell culture with amino acids and glucose and manufactured to meet cGMP manufacturing standards and QC specifications.

  • Designed to support modern manufacturing processes, Cell Boost 2 is chemically defined (CD), animal-derived component-free (ADCF), protein-free, and hydrolysate-free
  • For convenience, this cell culture supplement is available as a ready-to-use liquid format in addition to a dry powder format

Improve productivity and cell yield in fed-batch cultures by supplementing the cell culture medium to replace nutrients depleted during cell culture. HyClone Cell Boost 2 Supplement provides amino acids and glucose to support fed batch cultures in research and bioprocessing applications. It is available as a convenient ready-to-use liquid feed solution or as a dry powder feed to be hydrated before use.

This HyClone cell culture supplement has been developed for recombinant protein production with various cell lines including CHO cells and PER.C6 cells.

Supplement 100 to 200 ml/L, one or more times, starting during early- to mid-growth phase. This is expected to be day 2 to 4. Start the culture at a reduced volume according to anticipated feed volumes. Continue supplementation daily until the final volume is reached. It is recommended to monitor L- glutamine separately and supplement as needed.

Dry powdered medium milling equivalency study compares pin milling of powdered HyClone cell culture media and supplements at two manufacturing sites. Detailed physical, chemical, and biological data, including cell growth, are provided.

Specifikationer

Specifikationer

  • Pakn:
    100 g
  • Beskrivelse:
    Cell Boost™ 2
  • Størrelse:
    100 g
  • Form:
    Powder
  • Cat. No.:
    SH30596.01
  • Opbevaringstemperatur:
    2…8 °C

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.