Orders LinkContactUsLinkComponent
L-Glutamine, HyClone™
L-Glutamine 200 mM solution, concentration 29,2 mg/ml, 100 ml
Katalog # HYCLSH30034.01
L-Glutamine, HyClone™
Katalog # HYCLSH30034.01
CAS nummer:  
L-Glutamine, HyClone™
L-Glutamine 200 mM solution, concentration 29,2 mg/ml, 100 ml
CAS nummer:  
Price
150,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents
HyClone™ L-glutamine is used as a supplement in serum- and protein-free media. It supports the growth of cells that have high energy demands and synthesise large amounts of proteins and nucleic acids.
Specifikationer
Specifikationer
  • Pakn:
    100 ml
  • Beskrivelse:
    L-Glutamine 200 mM solution, concentration 29,2 mg/ml
Specifikationer