Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Zinc chromate

Zinc chromate

250 g
Katalog # A18178.30
Sundhed
Irriterende
Leverandør:   Thermo Fisher Scientific
Zinc chromate
Katalog # A18178.30
CAS nummer:  13530-65-9
Zinc chromate
Zinc chromate

250 g

CAS nummer:  13530-65-9
Price
916,00 kr
Log ind for tilgængelighed
1 * 250 g
Product Details & Documents

CAS No.: 13530-65-9 Molecular Formula: ZnCrO4 Formula Weight: 181.37 UN No.: UN3077 MDL No.: MFCD00036281

  • CAS nummer:13530-65-9
  • Formel:ZnCrO₄
  • MDL nummer:MFCD00036281
  • Molecular Weight:181.37 g/mol
  • Tæthed:3,4
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:236-878-9
  • ADR:9,III
Specifikationer

Specifikationer

  • Pakn:
    250 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.