Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Dibutyl sebacate ≥90%

Dibutyl sebacate ≥90%

Glass bottle for solids, 100 g
Katalog # A18231.22
Irriterende
Leverandør:   Thermo Fisher Scientific
Dibutyl sebacate ≥90%
Katalog # A18231.22
CAS nummer:  109-43-3
Dibutyl sebacate ≥90%
Dibutyl sebacate ≥90%

Glass bottle for solids, 100 g

CAS nummer:  109-43-3
Price
434,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Purity: 94% CAS No.: 109-43-3 Molecular Formula: CH3(CH2)3O2C(CH2)8CO2(CH2)3CH3 Formula Weight: 314.47 Flash Point: 178°(352°F) Refractive Index: 1.441 Melting Point: -11° Boiling Point: 344-345° Beilstein Reference: 1798308 MDL No.: MFCD00027218

Synonymer:  Sebacic acid dibutyl ester, 1,10-Decanedioic acid dibutyl ester, Dibutyl decanedioate
  • CAS nummer:109-43-3
  • Formel:C₁₈H₃₄O₄
  • MDL nummer:MFCD00027218
  • Molecular Weight:314.47 g/mol
  • Kogende Pt:344…345 °C
  • Smeltende Pt:–11 °C
  • Tæthed:0,94
  • Flash Pt:178 °C (352 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-672-5
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.