Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Dibutyl sebacate ≥90%

Dibutyl sebacate ≥90%

Glass bottle for solids, 500 g
Katalog # A18231.36
Irriterende
Leverandør:   Thermo Fisher Scientific
Dibutyl sebacate ≥90%
Katalog # A18231.36
CAS nummer:  109-43-3
Dibutyl sebacate ≥90%
Dibutyl sebacate ≥90%

Glass bottle for solids, 500 g

CAS nummer:  109-43-3
Price
1.370,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents

Purity: 94% CAS No.: 109-43-3 Molecular Formula: CH3(CH2)3O2C(CH2)8CO2(CH2)3CH3 Formula Weight: 314.47 Flash Point: 178°(352°F) Refractive Index: 1.441 Melting Point: -11° Boiling Point: 344-345° Beilstein Reference: 1798308 MDL No.: MFCD00027218

Synonymer:  Sebacic acid dibutyl ester, 1,10-Decanedioic acid dibutyl ester, Dibutyl decanedioate
  • CAS nummer:109-43-3
  • Formel:C₁₈H₃₄O₄
  • MDL nummer:MFCD00027218
  • Molecular Weight:314.47 g/mol
  • Kogende Pt:344…345 °C
  • Smeltende Pt:–11 °C
  • Tæthed:0,94
  • Flash Pt:178 °C (352 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-672-5
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g

Specifikationer