Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)
Semi-micro flow cell, RFID, 6 mm, 5 μl, 120 bar for G1315C/D, G1365C/D
Katalog # HEWLG1315-60025
Leverandør: AGILENT HEWLETT PACKARD
Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)
Katalog # HEWLG1315-60025
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Semi-micro flow cell, RFID, 6 mm, 5 μl, 120 bar for G1315C/D, G1365C/D
- Seq:09