Orders LinkContactUsLinkComponent
Drum fill gauge
Drum fill gauge
Pop op-tromle-påfyldningsmåler, forebygger overfyldning, 127 mm
Katalog # NEWPDRM245
Leverandør:   NEW PIG
Drum fill gauge
Katalog # NEWPDRM245
Leverandør:  NEW PIG
Drum fill gauge
Drum fill gauge
Pop op-tromle-påfyldningsmåler, forebygger overfyldning, 127 mm
Price
277,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These durable gauge helps prevent overfilling by popping up when drum is near capacity.
  • Corrosion-resistant polyethylene
  • Highly visible red and yellow offers easy indication
  • Avoid overflows and downtime by quickly checking a drum's liquid level
Specifikationer
  • Beskrivelse:
    Pop op-tromle-påfyldningsmåler, forebygger overfyldning
  • Længde:
    127 mm