Drum fill gauge
Pop op-tromle-påfyldningsmåler, forebygger overfyldning, 127 mm
Katalog # NEWPDRM245
Leverandør: NEW PIG
Gå til:
Product Details & Documents
These durable gauge helps prevent overfilling by popping up when drum is near capacity.
- Corrosion-resistant polyethylene
- Highly visible red and yellow offers easy indication
- Avoid overflows and downtime by quickly checking a drum's liquid level
Specifikationer
- Beskrivelse:Pop op-tromle-påfyldningsmåler, forebygger overfyldning
- Længde:127 mm