Insoles, blue
Insole for shoes, 47, blå
Catalog # 114-0268
Supplier: HONEYWELL SAFETY
Jump to:
Product Details & Documents
Blue non-woven fibres.
- Linear structure
- Perforated to encourage air circulation inside the shoe
- Impregnated with an anti-fungal and antibacterial agent
- Neutralises perspiration
- Combats germs
Specifications
- Skostørrelse:47
- Farve:blå
- Beskrivelse:Insole for shoes