Insoles, blue
Insole for shoes, 40, blå
Katalog # 114-0261
Leverandør: HONEYWELL SAFETY
Gå til:
Product Details & Documents
Blue non-woven fibres.
- Linear structure
- Perforated to encourage air circulation inside the shoe
- Impregnated with an anti-fungal and antibacterial agent
- Neutralises perspiration
- Combats germs
Specifikationer
- Skostørrelse:40
- Farve:blå
- Beskrivelse:Insole for shoes