Orders LinkContactUsLinkComponent
Insoles, blue
Insoles, blue
Insole for shoes, 39, blå
Katalog # 114-0260
Leverandør:   HONEYWELL SAFETY
Insoles, blue
Katalog # 114-0260
Leverandør:  HONEYWELL SAFETY
Insoles, blue
Insoles, blue
Insole for shoes, 39, blå
Price
921,00 kr
Log ind for tilgængelighed
1 * 10 Par
Product Details & Documents
Blue non-woven fibres.
  • Linear structure
  • Perforated to encourage air circulation inside the shoe
  • Impregnated with an anti-fungal and antibacterial agent
  • Neutralises perspiration
  • Combats germs
Specifikationer
  • Skostørrelse:
    39
  • Farve:
    blå
  • Beskrivelse:
    Insole for shoes