MonoBox, TripleBox
Monobox med 1 Tempshell 22 °C, 390×290×230 mm
Katalog # 478-9930
Leverandør: DELTA T
Gå til:
Product Details & Documents
These extra strong insulated boxes can be used for delivery. Made from Neopor® the boxes consist of 2 different parts which can be fitted together to create different volumes.
- Eliminates the cost of expensive refrigeration transportation
- Maintains the required temperature
- Reusable
The MonoTripleBox can be used to transport test tubes or other products.
Specifikationer
- Beskrivelse:Monobox med 1 Tempshell 22 °C
- Seq:03
- B×D×H:390×290×230 mm



