MonoBox, TripleBox
2-dels TripleBox med 1 Tempshell 22 °C og 2 Tempframes, 390×290×350 mm
478-9936
: DELTA T
These extra strong insulated boxes can be used for delivery. Made from Neopor® the boxes consist of 2 different parts which can be fitted together to create different volumes.
- Eliminates the cost of expensive refrigeration transportation
- Maintains the required temperature
- Reusable
The MonoTripleBox can be used to transport test tubes or other products.
- Beskrivelse:2-dels TripleBox med 1 Tempshell 22 °C og 2 Tempframes
- Seq:07
- B×D×H:390×290×350 mm