Orders LinkContactUsLinkComponent
MonoBox, TripleBox
2-dels TripleBox med 1 Tempshell 22 °C og 2 Tempframes, 390×290×350 mm
  478-9936
 :   DELTA T
MonoBox, TripleBox
  478-9936
 :  DELTA T
MonoBox, TripleBox
MonoBox, TripleBox
2-dels TripleBox med 1 Tempshell 22 °C og 2 Tempframes, 390×290×350 mm
3.230,00 kr
1 * 1 Items
These extra strong insulated boxes can be used for delivery. Made from Neopor® the boxes consist of 2 different parts which can be fitted together to create different volumes.
  • Eliminates the cost of expensive refrigeration transportation
  • Maintains the required temperature
  • Reusable
The MonoTripleBox can be used to transport test tubes or other products.
  • Beskrivelse:
    2-dels TripleBox med 1 Tempshell 22 °C og 2 Tempframes
  • Seq:
    07
  • B×D×H:
    390×290×350 mm