MonoBox, TripleBox
2-dels TripleBox med 1 Tempshell 4 °C og 2 Tempframes, 390×290×350 mm
Catalog # 478-9935
Supplier: DELTA T
Jump to:
Product Details & Documents
These extra strong insulated boxes can be used for delivery. Made from Neopor® the boxes consist of 2 different parts which can be fitted together to create different volumes.
- Eliminates the cost of expensive refrigeration transportation
- Maintains the required temperature
- Reusable
The MonoTripleBox can be used to transport test tubes or other products.
Specifications
- Beskrivelse:2-dels TripleBox med 1 Tempshell 4 °C og 2 Tempframes
- Seq:06
- B×D×H:390×290×350 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



