Sample vials, clear, with screw thread, KIMBLE®
10,00 ml, 20×55 mm, klar, Yes
Katalog # SCERC40552000B0B2
Leverandør: DWK Life Sciences
Sample vials, clear, with screw thread, KIMBLE®
Katalog # SCERC40552000B0B2
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
Screw threaded sample vials made from soda-lime glass, with white PE caps unattached.
Documents
Specifikationer
- Kasket inkluderet:Yes
- Diameter×H:20×55 mm
- Kapacitet:10,00 ml
- Farve:klar
- Seq:03