ISA opløsninger
Ionstyrke justering for Klorid ion selektiv elektrode, i en flaske, Ion selective electrode reagents, Bottle, 475 mL
Ionic strength adjustors are used for analysis with ion selective electrodes, and special reagents are used with some ion selective electrodes.
- Ionic strength adjuster is added to all samples and standards to create a uniform background ionic strength, which provides more reproducible measurements
- Ammonia pH-adjusting ISA and low-level ammonia pH-adjusting ISA both include a pH-indicating blue dye for instant verification of correct pH in ammonia samples and standards with no pH measurement required
- Ammonia pH-adjusting ISA prevents complexation of ammonia and inhibits formation of metal hydroxides, allowing determination of total ammonia in the sample and increasing the accuracy of results
- Low-level ammonia pH-adjusting ISA is ideal for ammonia samples with a concentration of 10 ppm and below that do not contain metal interferences
Concentration : 10 ppm
For Use With (Equipment) : Ion selective electrode
For Use With (Application) : Ionic strength adjustors used for analysis with ion selective electrodes and special reagents used with some ion selective electrodes
- Pakning:Bottle
- Pakn:475 mL
- Beskrivelse:Ionstyrke justering for Klorid ion selektiv elektrode, i en flaske
- Model:Ion selective electrode reagents



