Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
ISA opløsninger

ISA opløsninger

Ionstyrke justering for natrium ion selektiv elektrode, i en flaske, Ion selective electrode reagents, Bottle, 475 mL
Katalog # 662-0745
Leverandør:   Thermo Orion
ISA opløsninger
Katalog # 662-0745
Leverandør:  Thermo Orion
CAS nummer:  
ISA opløsninger
ISA opløsninger

Ionstyrke justering for natrium ion selektiv elektrode, i en flaske, Ion selective electrode reagents, Bottle, 475 mL

CAS nummer:  
Price
819,00 kr
Log ind for tilgængelighed
1 * 475 mL
Product Details & Documents

Ionic strength adjustors are used for analysis with ion selective electrodes, and special reagents are used with some ion selective electrodes.

  • Ionic strength adjuster is added to all samples and standards to create a uniform background ionic strength, which provides more reproducible measurements
  • Ammonia pH-adjusting ISA and low-level ammonia pH-adjusting ISA both include a pH-indicating blue dye for instant verification of correct pH in ammonia samples and standards with no pH measurement required
  • Ammonia pH-adjusting ISA prevents complexation of ammonia and inhibits formation of metal hydroxides, allowing determination of total ammonia in the sample and increasing the accuracy of results
  • Low-level ammonia pH-adjusting ISA is ideal for ammonia samples with a concentration of 10 ppm and below that do not contain metal interferences

Concentration : 10 ppm

For Use With (Equipment) : Ion selective electrode
For Use With (Application) : Ionic strength adjustors used for analysis with ion selective electrodes and special reagents used with some ion selective electrodes

Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    475 mL
  • Beskrivelse:
    Ionstyrke justering for natrium ion selektiv elektrode, i en flaske
  • Model:
    Ion selective electrode reagents

Specifikationer