Accessories
Waterproof kit 7,5 L, G 788 (inc CD), PG 8535
Katalog # MIEL5168033
Gå til:
Discontinued
Product Details & Documents
- For lower and upper baskets
- With 7 x 7 mm perforations
- Stainless steel
Specifikationer
- Beskrivelse:Waterproof kit 7,5 L
- Seq:49
- Til:G 788 (inc CD), PG 8535