Racks for pipette boxes
Pipettereol, 4 sterilisationspipette-kasser, Rustfrit stål, 200×250×225 mm
Katalog # GLWAP04
Leverandør: GLW
Gå til:
Product Details & Documents
These stainless steel racks are suitable for the storage of four or six GLW pipette boxes (A30, A40, A45 / S30, S40, S45).
Bestillingsoplysninger: Pipette boxes are not included.
Specifikationer
- Til:4 sterilisationspipette-kasser
- Beskrivelse:Pipettereol
- Materiale:Rustfrit stål
- B×D×H:200×250×225 mm