Slide Boxes
sort, PS, 50, 172×83×31 mm
Katalog # GLWAK50S
Leverandør: GLW
Gå til:
Product Details & Documents
High quality slide storage boxes for microscopy slides.
Used for storing, transporting and freezing in science, industry and biotechnology.
Specifikationer
- Antal objektglas:50
- Farve:sort
- Materiale:PS
- Seq:05
- B×D×H:172×83×31 mm
Accessories
Produktinformation
- Katalog #FarveMaterialeTilgængelighedPrice
- Divider for slides and coverslips of special sizesDivider for slides and coverslips of special sizesGLWAZK20gråPSLog ind for tilgængelighed1 * 1 Items - 24,40 kr
Produktinformation
- Katalog #FarveMaterialeTilgængelighedPrice
- Divider for slides and coverslips of special sizesDivider for slides and coverslips of special sizesGLWAZK15hvidPSLog ind for tilgængelighed1 * 1 Each - 51,70 kr