Orders LinkContactUsLinkComponent
Sample bottles, glass
Sample bottles, glass
1000 ml, Prøveudtagningsflaske, glas, 1000 ml, GL 45
Katalog # BURK5314-0100
Leverandør:   BURKLE
Sample bottles, glass
Katalog # BURK5314-0100
Leverandør:  BURKLE
Sample bottles, glass
Sample bottles, glass
1000 ml, Prøveudtagningsflaske, glas, 1000 ml, GL 45
Price
333,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Specifikationer
  • Gevind:
    GL 45
  • Kapacitet:
    1000 ml
  • Beskrivelse:
    Prøveudtagningsflaske, glas, 1000 ml
  • Seq:
    03