[EN]PCB REGULATION WITH HEAT SENSOR 1 * 1 Items
Katalog # UVPA75-0047-02
Leverandør: UVP ULTRA VIOLET PRODUCTS
[EN]PCB REGULATION WITH HEAT SENSOR 1 * 1 Items
Katalog # UVPA75-0047-02
Leverandør: UVP ULTRA VIOLET PRODUCTS
Frequently Bought Together
VWR® PureStep, Multi-Layer Adhesive Floor Mats
Leverandør: VWR Collection
Acetone ≥99%, TECHNICAL
Leverandør: VWR Chemicals
KABEL H11 1 * 1 Items
Leverandør: IKA
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverandør: VWR Chemicals
Lithium Polymer Rechargable Battery
Leverandør: EPPENDORF
Acetonitril, LiChrosolv®, hypergrade for LC-MS, Supelco®
Leverandør: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Leverandør: VWR Chemicals
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Leverandør: HIRSCHMANN
Accessories for accu-jet® pro
Leverandør: Brand
Combitips® advanced PCR Clean - Positive Displacement Pipette Tips
Leverandør: EPPENDORF
Vand, LiChrosolv® for chromatography, for LC-MS, Supelco®
Leverandør: Merck
Accessories for electronic dispensers, Multipette E3/E3x
Leverandør: EPPENDORF
VWR®, Accessories for Mechanical Pipettes
Leverandør: VWR Collection
Accessories for HandyStep® electronic
Leverandør: Brand
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverandør: VWR Chemicals
Vand, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverandør: VWR Chemicals