Vippemåler
5 ml
BRND43055
: Brand
Dispenser head made of borosilicate glass 3.3, with inscriptions in green enamel.
- Calibrated to deliver (TD, Ex)
- Excellent chemical resistance
- Excellent good temperature resistance
Automatic dispenser particularly suited for dispensing suspensions.
Ordering information: Reservoir bottles available on request.
Documents
Brand Volumetric Measurement in the Laboratory
Produktlitteratur
- Kapacitet:5 ml
- Seq:01