Orders LinkContactUsLinkComponent
Ammoniumnitrat ≥98.5%, TECHNICAL
Plastic bucket, 25 kg
  21277.460
Ammoniumnitrat ≥98.5%, TECHNICAL
  21277.460
 :  6484-52-2
Ammoniumnitrat ≥98.5%, TECHNICAL
Ammoniumnitrat ≥98.5%, TECHNICAL
Plastic bucket, 25 kg
 :  6484-52-2
3.050,00 kr
1 * 25 kg
  •  :6484-52-2
  •  :NH₄NO₃
  •  :MFCD00011425
  •  :80.04 g/mol
  •  :13,00538
  •  :230 °C (1013 hPa)
  •  :169 °C
  •  :1,73 g/cm³ (20 °C)
  •  :Stue temperatur
  •  :229-347-8
  •  :1942
  •  :5.1,III
  • Assay (calculated on anhydrous):
    Min. 98.5 %
  • Pakning:
    Plastic bucket
  • Pakn:
    25 kg