Orders LinkContactUsLinkComponent
PIG® BLUE absorbent socks
PIG® BLUE absorbent socks
Absorbent socks, PP skin/vermiculate filler, 56 L, 8 strømper/æske, 80×3000 mm
  NEWPPIG202
 :   NEW PIG
PIG® BLUE absorbent socks
  NEWPPIG202
 :  NEW PIG
PIG® BLUE absorbent socks
PIG® BLUE absorbent socks
Absorbent socks, PP skin/vermiculate filler, 56 L, 8 strømper/æske, 80×3000 mm
1.170,00 kr
1 * 8 Items
Surround spills to prevent spreading. Extra-fine vermiculite filler pulls in liquid and hugs floors to stay in place.
  • Heaviest sock, features extra-dense construction to hug floors and stay in place whilst containing spills
  • Fine-grade vermiculite filler quickly absorbs leaks, drips and spills; great for spill response or absorbing machine leaks
  • Stitch-bonded, polypropylene skin resists bursting; reduces dust and holds in liquid, even when saturated
  • Absorbs and retains oils, coolants, solvents and water
Ideal for diking and damming liquids. Can be air-dried for reuse when soaked with water.
  • Beskrivelse:
    Absorbent socks, PP skin/vermiculate filler
  • Seq:
    05
  • Sugeevne:
    56 L
  • Pakket:
    8 strømper/æske
  • Ø×L:
    80×3000 mm