Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

PIG® BLUE absorbent socks

Absorbent socks, PP skin/vermiculate filler, 78,1 L, 55 strømper/æske, 80×610 mm
Katalog # NEWPPIG201
Leverandør:   NEW PIG
PIG® BLUE absorbent socks
Katalog # NEWPPIG201
Leverandør:  NEW PIG
PIG® BLUE absorbent socks
PIG® BLUE absorbent socks

Absorbent socks, PP skin/vermiculate filler, 78,1 L, 55 strømper/æske, 80×610 mm

Price
1.440,00 kr
Log ind for tilgængelighed
1 * 55 Items
Product Details & Documents

Surround spills to prevent spreading. Extra-fine vermiculite filler pulls in liquid and hugs floors to stay in place.

  • Heaviest sock, features extra-dense construction to hug floors and stay in place whilst containing spills
  • Fine-grade vermiculite filler quickly absorbs leaks, drips and spills; great for spill response or absorbing machine leaks
  • Stitch-bonded, polypropylene skin resists bursting; reduces dust and holds in liquid, even when saturated
  • Absorbs and retains oils, coolants, solvents and water

Ideal for diking and damming liquids. Can be air-dried for reuse when soaked with water.

Specifikationer
  • Beskrivelse:
    Absorbent socks, PP skin/vermiculate filler
  • Seq:
    02
  • Sugeevne:
    78,1 L
  • Pakket:
    55 strømper/æske
  • Ø×L:
    80×610 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.