Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

(±)-Linalool ≥96.0% (by GC)

Glass bottle, 25 mL
Katalog # TCIAL0048-25ML
Irriterende
Sundhed
Leverandør:   TCI
(±)-Linalool ≥96.0% (by GC)
Katalog # TCIAL0048-25ML
Leverandør:  TCI
CAS nummer:  78-70-6
(±)-Linalool ≥96.0% (by GC)
(±)-Linalool ≥96.0% (by GC)

Glass bottle, 25 mL

CAS nummer:  78-70-6
Price
132,00 kr
Log ind for tilgængelighed
1 * 25 mL
Product Details & Documents
Synonymer:  (±)-3,7-Dimethyl-1,6-octadien-3-ol, 3,7-Dimethylocta-1,6-dien-3-ol
  • CAS nummer:78-70-6
  • Formel:C₁₀H₁₈O
  • MDL nummer:MFCD00008906
  • Molecular Weight:154.25 g/mol
  • Merck Index:14,05495
  • Kogende Pt:200°C
  • Tæthed:0,87 g/cm³ (20 °C)
  • Flash Pt:71°C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:201-134-4
Specifikationer

Specifikationstestresultater

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 96.0 %

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    25 mL

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.