Calibration weights, OIML, class M2
10000 g, 1600 ± mg, Blok uden certifikat
Katalog # SARBYCW7169-AC-00
Leverandør: Sartorius Balances
Gå til:
Product Details & Documents
Weights are used for control of inspection, measuring and test equipment.
Proof Line series, OIML Class M2 knob weights are manufactured from brass, precision-lathed, fine finish with the density of 8,400 kg/m3 and the block weights are made up of cast iron, gray cast, black lacquered finish with the density of 7,200 kg/m3.
: Compliance with OIML R111-1.
Specifikationer
- Vægt:10000 g
- Seq:08
- Tolerance:1600 ± mg
- Model:Blok uden certifikat



