Calibration weights, OIML, class M2
2000 g, 300 ± mg, Lod med DKD-certifikat
SARBYCW626-AC-02
Weights are used for control of inspection, measuring and test equipment.
Proof Line series, OIML Class M2 knob weights are manufactured from brass, precision-lathed, fine finish with the density of 8,400 kg/m3 and the block weights are made up of cast iron, gray cast, black lacquered finish with the density of 7,200 kg/m3.
: Compliance with OIML R111-1.
- Vægt:2000 g
- Seq:07
- Tolerance:300 ± mg
- Model:Lod med DKD-certifikat