Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Calibration weights, OIML, class M2

Calibration weights, OIML, class M2

500 g, 80 ± mg, Lod uden certifikat
Katalog # SARBYCW556-AC-00
Leverandør:   Sartorius Balances
Calibration weights, OIML, class M2
Katalog # SARBYCW556-AC-00
Leverandør:  Sartorius Balances
Calibration weights, OIML, class M2
Calibration weights, OIML, class M2

500 g, 80 ± mg, Lod uden certifikat

Price
490,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Weights are used for control of inspection, measuring and test equipment.

Proof Line series, OIML Class M2 knob weights are manufactured from brass, precision-lathed, fine finish with the density of 8,400 kg/m3 and the block weights are made up of cast iron, gray cast, black lacquered finish with the density of 7,200 kg/m3.

Certifications: Compliance with OIML R111-1.

Specifikationer
  • Vægt:
    500 g
  • Seq:
    03
  • Tolerance:
    80 ± mg
  • Model:
    Lod uden certifikat
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.