Staining jar and dish
Staining jar, with 2 lids, TPX
Katalog # DELT19353
Leverandør: DELTALAB
Gå til:
Product Details & Documents
Staining dish made of polypropylene and jar made of TPX.
- Supplied with two lids
- First lid provides evaporation free storage of staining liquid when the dish is not in use
- Second lid includes a slot to allow staining dish insertion
Documents
Certifikater
Søg efter partinummerSpecifikationer
- Beskrivelse:Staining jar, with 2 lids, TPX
- Seq:01