Orders LinkContactUsLinkComponent
Staining jar and dish
Staining jar and dish
Staining jar, with 2 lids, TPX
Katalog # DELT19353
Leverandør:   DELTALAB
Staining jar and dish
Katalog # DELT19353
Leverandør:  DELTALAB
Staining jar and dish
Staining jar, with 2 lids, TPX
Price
679,00 kr
Log ind for tilgængelighed
1 * 4 Items
Product Details & Documents
Staining dish made of polypropylene and jar made of TPX.
  • Supplied with two lids
  • First lid provides evaporation free storage of staining liquid when the dish is not in use
  • Second lid includes a slot to allow staining dish insertion
Documents
Specifikationer
  • Beskrivelse:
    Staining jar, with 2 lids, TPX
  • Seq:
    01