L(-)-Carnitine
10 Vial
Catalog # CAMBNSK-B
Supplier: CAMBRIDGE ISOTOPE LABS
Jump to:
Product Details & Documents
Synonyms: Levocarnitine, L-Carnitine, Vitamin BT, (R)-3-Hydroxy-4-(trimethylammonio)butanoate, R)-(-)-3-Hydroxy-4-(trimethylammonio)butyrate
- CAS Number:541-15-1
- Formula:C₇H₁₅NO₃
- MDL Number:MFCD00038747
- Molecular Weight:161.20 g/mol
- EINECS:208-768-0
Specifications
Specifications
- Pakn:10 Vial



