Orders LinkContactUsLinkComponent
Cover glasses
Cover glasses, square, without cords and blisters, 25×40 mm, #1, Borosilikatglas
  KNITVD12540Y100A
Cover glasses
  KNITVD12540Y100A
 :  
Cover glasses
Cover glasses, square, without cords and blisters, 25×40 mm, #1, Borosilikatglas
Borosilicate glass of hydrolytic class 1 cover glasses with precise cutting.
 : ISO 8255/1
 : 100 pieces per box - 1000 pieces per carton.
  • Antal:
    #1
  • Beskrivelse:
    Cover glasses, square, without cords and blisters
  • L×B:
    25×40 mm
  • Materiale:
    Borosilikatglas