Cover glasses
Cover glasses, square, without cords and blisters, 25×40 mm, #1, Borosilikatglas
KNITVD12540Y100A
Borosilicate glass of hydrolytic class 1 cover glasses with precise cutting.
: ISO 8255/1
: 100 pieces per box - 1000 pieces per carton.
- Antal:#1
- Beskrivelse:Cover glasses, square, without cords and blisters
- L×B:25×40 mm
- Materiale:Borosilikatglas