Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Cover glasses

Cover glasses, 0,13 - 0,16 mm, 12 mm, Glas
  BELC1943-10012A
Marketsource
Cover glasses
  BELC1943-10012A
Marketsource
Cover glasses
Cover glasses

Cover glasses, 0,13 - 0,16 mm, 12 mm, Glas

 
173,60 kr
   
1 * 100 Items
 

Round cover slips made of glass.

 
  • Ø:
    12 mm
  • Beskrivelse:
    Cover glasses
  • Materiale:
    Glas
  • Seq:
    003
  • Tykkelse:
    0,13 - 0,16 mm