Sodium Hydrolysate Buffer 5 1 * 2 L
Katalog # WAPR80-2038-09
Leverandør: Biochrom
Frequently Bought Together
VWR® AVS TITRINORM, bufferopløsninger, 20 °C
Leverandør: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Leverandør: Merck
Acetone ≥99%, TECHNICAL
Leverandør: VWR Chemicals
pH-teststrimler, MQuant®, Supelco®
Leverandør: Merck
Kaliumchlorid 3 mol/l (3 N) in aqueous solution, Supelco®
Leverandør: Merck
VWR®, Conductivity standard solutions
Leverandør: VWR Chemicals
Sodium Regeneration Buffer 6 1 * 1 L
Leverandør: Biochrom
Ultra Ninhydrin Kit (4x2 L) 1 * 1 Each
Leverandør: Biochrom
BUFFER SOL. VALVE 1 * 1 Items
Leverandør: Biochrom
Sodium Hydrolysate Buffer 1 1 * 2 L
Leverandør: Biochrom
Sodium Hydro/Oxid Buffer 2 1 * 2 L
Leverandør: Biochrom
Ultra Ninhydrin Reagent Kit 1 * 2 L
Leverandør: Biochrom