Staining dishes, WHEATON®
Handles for staining racks, - mm
Katalog # 631-2505
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
Soda-lime glass and stainless steel.
- Removable glass slide rack has an open bottom to facilitate rapid immersion and drainage, reducing carry-over
Standard for manual staining procedures. The rack holds 10 single slides, 19 slides arranged alternately straight across and diagonally, or 20 slides back to back of standard size 75×25 mm, 75×38 mm and 75×51 mm.
Documents
WHEA900200_TDS_EN
Datablad
Specifikationer
- Beskrivelse:Handles for staining racks
- B×D×H:- mm