Orders LinkContactUsLinkComponent
Staining dishes, WHEATON®
Staining dishes, WHEATON®
Staining dish with cover, 95×76×64 mm
Katalog # 631-2503
Leverandør:   DWK Life Sciences
Staining dishes, WHEATON®
Katalog # 631-2503
Leverandør:  DWK Life Sciences
Staining dishes, WHEATON®
Staining dishes, WHEATON®
Staining dish with cover, 95×76×64 mm
Price
2.980,00 kr
Log ind for tilgængelighed
1 * 3 Items
Product Details & Documents
Soda-lime glass and stainless steel.
  • Removable glass slide rack has an open bottom to facilitate rapid immersion and drainage, reducing carry-over
Standard for manual staining procedures. The rack holds 10 single slides, 19 slides arranged alternately straight across and diagonally, or 20 slides back to back of standard size 75×25 mm, 75×38 mm and 75×51 mm.
Documents
Specifikationer
  • Beskrivelse:
    Staining dish with cover
  • B×D×H:
    95×76×64 mm