Pipette tips, ratiolab®
Dr. Lange, 100 - 1000 µl, -, Transparent, Dr. Lange-pipetter, 1.000 stk./pose
Catalog # RATI2500130
Supplier: RATIOLAB
Jump to:
Product Details & Documents
ratiolab® pipette tips are ideal for use with all commercially available pipettes.
- Made of high quality PP
- Tight fit due to the precisely fitting cone
- Guaranteed dust-free due to special ionization treatment
- Excellent transparency due to high quality surfaces
- Minimal residual wetting due to controlled quality of raw materials
- Excellent volume accuracy due to precise tip orifice
- MultiTray® refill system is space- and cost-saving and clearly reduces the amount of waste
MultiTray®: Dust-proof refill pack for the Multibox® plus and the Multibox® ultra.
Specifications
- Pakket:1.000 stk./pose
- Steril:-
- Volumen:100 - 1000 µl
- Seq:50
- Spidstype:Dr. Lange
- Spits farge:Transparent
- Til brug med:Dr. Lange-pipetter