Orders LinkContactUsLinkComponent
Calibrated Baumé hydrometers
10 - 20, 0,1, 300 mm
  ALLA0101FC020/20QP
Calibrated Baumé hydrometers
  ALLA0101FC020/20QP
Calibrated Baumé hydrometers
Calibrated Baumé hydrometers
10 - 20, 0,1, 300 mm
506,00 kr
1 * 1 Items
Baumé hydrometers calibrated at 20 °C.
  • Længde:
    300 mm
  • Seq:
    04
  • Inddeling:
    0,1
  • Område:
    10 - 20