PIG® spill caddy
Refill haz-mat spill caddy, 47,4 L
Katalog # 121-0517
Leverandør: NEW PIG
Gå til:
Product Details & Documents
The mobile PIG® spill caddy for fast spill response or daily leak maintenance.
- High-visibility yellow container is easy to spot when you need it
- Contents are compartmentalised for easy access
- Adjustable shelves
- 20 cm wheels for easy mobility
- Double-wall polyethylene cart for durability
Container can be locked with a padlock (not included) to prevent pilferage.
Advarsel: To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of your chemicals with PIG® HAZ-MAT products prior to purchase.
Specifikationer
- Beskrivelse:Refill haz-mat spill caddy
- Seq:04
- Sugeevne:47,4 L