Disposal Poly Bags
Små poser, PE, 762×457 mm
121-0503
: NEW PIG
These chemical resistant bags are an ideal way to handle hazardous or contaminated waste while providing temporary storage or disposal in a safe manner. Polyethylene.
- Black warning 'Hazardous Material Handle With Care' on bright yellow
- Ideal for lining drums
- Includes self-locking ties
- Beskrivelse:Små poser
- L×B:762×457 mm
- Materiale:PE