Orders LinkContactUsLinkComponent
Disposal Poly Bags
Små poser, PE, 762×457 mm
  121-0503
 :   NEW PIG
Disposal Poly Bags
  121-0503
 :  NEW PIG
Disposal Poly Bags
Disposal Poly Bags
Små poser, PE, 762×457 mm
619,00 kr
1 * 50 Items
These chemical resistant bags are an ideal way to handle hazardous or contaminated waste while providing temporary storage or disposal in a safe manner. Polyethylene.
  • Black warning 'Hazardous Material Handle With Care' on bright yellow
  • Ideal for lining drums
  • Includes self-locking ties
  • Beskrivelse:
    Små poser
  • L×B:
    762×457 mm
  • Materiale:
    PE