Disposal Poly Bags
Store poser, PE, 1524×914 mm
Katalog # 121-0502
Leverandør: NEW PIG
Gå til:
Product Details & Documents
These chemical resistant bags are an ideal way to handle hazardous or contaminated waste while providing temporary storage or disposal in a safe manner. Polyethylene.
- Black warning 'Hazardous Material Handle With Care' on bright yellow
- Ideal for lining drums
- Includes self-locking ties
Specifikationer
- Beskrivelse:Store poser
- L×B:1524×914 mm
- Materiale:PE