Orders LinkContactUsLinkComponent
Disposal Poly Bags
Store poser, PE, 1524×914 mm
Katalog # 121-0502
Leverandør:   NEW PIG
Disposal Poly Bags
Katalog # 121-0502
Leverandør:  NEW PIG
Disposal Poly Bags
Disposal Poly Bags
Store poser, PE, 1524×914 mm
Price
946,00 kr
Log ind for tilgængelighed
1 * 25 Items
Product Details & Documents
These chemical resistant bags are an ideal way to handle hazardous or contaminated waste while providing temporary storage or disposal in a safe manner. Polyethylene.
  • Black warning 'Hazardous Material Handle With Care' on bright yellow
  • Ideal for lining drums
  • Includes self-locking ties
Specifikationer
  • Beskrivelse:
    Store poser
  • L×B:
    1524×914 mm
  • Materiale:
    PE