Orders LinkContactUsLinkComponent
Accessories for DOSE IT
Tube collar used as a weight for the aspiration tube or as a clamp spacer for the dispensing tube, 4 − 6 mm
Katalog # 710-1041
Leverandør:   INTEGRA Biosciences
Accessories for DOSE IT
Katalog # 710-1041
Leverandør:  INTEGRA Biosciences
Accessories for DOSE IT
Accessories for DOSE IT
Tube collar used as a weight for the aspiration tube or as a clamp spacer for the dispensing tube, 4 − 6 mm
Price
658,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Tube collar used as a weight for the aspiration tube or as a clamp spacer for the dispensing tube
  • Ø indv.:
    4 − 6 mm