Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
VWR®, Urglas

VWR®, Urglas

Beaker covers/watch glasses, PTFE, 200 mm
Katalog # 216-0455
Leverandør:   VWR Collection
VWR®, Urglas
Katalog # 216-0455
Leverandør:  VWR Collection
VWR®, Urglas
VWR®, Urglas

Beaker covers/watch glasses, PTFE, 200 mm

Price
389,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

PTFE.

  • Excellent chemical resistance
  • Temperature resistant from –200 to 280 °C
  • Exceptionally smooth surfaces to prevent contamination

Ideal for covering PTFE beakers when decomposing and evaporating, can also be used as evaporating dishes. Extremely robust.

Specifikationer
  • Ø:
    200 mm
  • Beskrivelse:
    Beaker covers/watch glasses, PTFE
  • Seq:
    11